Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKRN4P
Entrez Gene ID
395500
Gene Name
makorin ring finger protein 4, pseudogene
Gene Aliases
MKRN1
Chromosome Location
Chr.1: 56777619 - 56801776 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204742.2 NP_990073.1 2 - 3 366 86
XM_015284432.2 XP_015139918.1 2 - 3 366 86
GenBank mRNA AF192787.1 - 2 - 3 300 86

View Full Product Details