Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKRN4P
Entrez Gene ID
395500
Gene Name
makorin ring finger protein 4, pseudogene
Gene Aliases
MKRN1
Chromosome Location
Chr.1: 56777619 - 56801776 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204742.2 NP_990073.1 5 - 6 1025 94
XM_015284432.2 XP_015139918.1 5 - 6 1025 94
GenBank mRNA AF192787.1 - 5 - 6 959 94

View Full Product Details