Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP2
Entrez Gene ID
395546
Gene Name
secreted frizzled related protein 2
Gene Aliases
SFRP-2
Chromosome Location
Chr.4: 20063707 - 20067350 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204773.1 NP_990104.1 2 - 3 597 121
GenBank mRNA AF218056.1 - 2 - 3 597 121

View Full Product Details