Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGST1
Entrez Gene ID
418178
Gene Name
microsomal glutathione S-transferase 1
Gene Aliases
-
Chromosome Location
Chr.1: 63277208 - 63283946 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135550.1 NP_001129022.1 2 - 3 230 129
XM_015290135.2 XP_015145621.1 2 - 3 241 129
XM_015290146.2 XP_015145632.1 2 - 3 234 129
XM_025148902.1 XP_025004670.1 2 - 3 227 129
XM_025148903.1 XP_025004671.1 2 - 3 183 129
XM_025148905.1 XP_025004673.1 2 - 3 423 129
XM_025148906.1 XP_025004674.1 2 - 3 262 129
XM_025148908.1 XP_025004676.1 2 - 3 414 129
GenBank mRNA EF585397.1 - 2 - 3 216 129
EF585398.1 - 2 - 3 230 129
EU309478.1 - 2 - 3 218 129
EU309479.1 - 2 - 3 228 129

View Full Product Details