Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NANOG
Entrez Gene ID
100272166
Gene Name
Nanog homeobox
Gene Aliases
-
Chromosome Location
Chr.1: 75593243 - 75596024 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146142.1 NP_001139614.1 1 - 2 157 68
GenBank mRNA DQ867025.1 - 1 - 2 157 68

View Full Product Details