Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIR
Entrez Gene ID
547693
Gene Name
ferredoxin:sulfite reductase precursor
Gene Aliases
DCP68, GmSiR
Chromosome Location
Chr.11: 7051681 - 7056161 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY017473.1 - 3 - 4 1323 109

View Full Product Details