Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDR12
Entrez Gene ID
732607
Gene Name
PDR-like ABC-transporter
Gene Aliases
-
Chromosome Location
Chr.3: 40242289 - 40250187 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250768.1 NP_001237697.1 2 - 3 551 94
GenBank mRNA AM261476.1 - 2 - 3 551 94

View Full Product Details