Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BMY2
- Entrez Gene ID
-
547683
- Gene Name
- beta-amylase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 12701241 - 12704431 on Build Glycine_max_v2.1
- Gene Symbol
- LOC100818063
- Entrez Gene ID
-
100818063
- Gene Name
- beta-amylase 3, chloroplastic-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 5167564 - 5171159 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK245644.1 |
- |
1 - 2 |
558 |
95 |
View Full Product Details