Gene Transcripts
- Gene Symbol
- DOCK3
- Entrez Gene ID
- 1795
- Gene Name
- dedicator of cytokinesis 3
- Gene Aliases
- MOCA, PBP
- Chromosome Location
- Chr. 3: 50674969 - 51384198 on Build GRCh38
- UniGene ID
- Hs.476284
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004947.4 |
NP_004938.1
|
5, 6 |
332 |
|
XM_005264914.3 |
XP_005264971.1
|
5, 6 |
604 |
|
XM_005264915.3 |
XP_005264972.1
|
5, 6 |
604 |
|
XM_005264916.3 |
XP_005264973.1
|
5, 6 |
382 |
|
XM_005264917.3 |
XP_005264974.1
|
5, 6 |
604 |
|
XM_005264918.3 |
XP_005264975.1
|
5, 6 |
381 |
|
XM_006713008.3 |
XP_006713071.1
|
5, 6 |
604 |
|
XM_006713009.3 |
XP_006713072.1
|
5, 6 |
604 |
|
XM_006713010.3 |
XP_006713073.1
|
5, 6 |
367 |
|
XM_017005825.1 |
XP_016861314.1
|
5, 6 |
383 |
|
XM_017005826.1 |
XP_016861315.1
|
5, 6 |
373 |
|
GenBank mRNA |
AY145303.1 |
- |
5, 6 |
309 |
|
AY254099.1 |
- |
5, 6 |
332 |
|
View Full Product Details