Gene Transcripts

Gene Symbol
HOXC11
Entrez Gene ID
3227
Gene Name
homeobox C11
Gene Aliases
HOX3H
Chromosome Location
Chr. 12: 53973126 - 53976419 on Build GRCh38
UniGene ID
Hs.127562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014212.3 NP_055027.1 2 932
GenBank mRNA AJ000041.1 - 2 860
AK291946.1 - 2 930
BC001543.1 - 2 913 3462682
X99630.1 - 1 96

View Full Product Details