Gene Transcripts
- Gene Symbol
- PSMB1
- Entrez Gene ID
- 5689
- Gene Name
- proteasome subunit beta 1
- Gene Aliases
- HC5, PMSB1, PSC5
- Chromosome Location
- Chr. 6: 170535116 - 170553329 on Build GRCh38
- UniGene ID
- Hs.352768
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002793.3 |
NP_002784.1
|
1, 2 |
201 |
|
GenBank mRNA |
AB209078.1 |
- |
1, 2 |
131 |
|
AB451312.1 |
- |
1, 2 |
114 |
|
AB451442.1 |
- |
1, 2 |
114 |
|
AK058050.1 |
- |
1, 2 |
171 |
|
AK223193.1 |
- |
1, 2 |
152 |
|
BC000508.2 |
- |
1, 2 |
152 |
2822268 |
BC020807.1 |
- |
1, 2 |
152 |
4281426 |
BP369175.1 |
- |
1, 2 |
213 |
|
BT019720.1 |
- |
1, 2 |
114 |
|
D00761.1 |
- |
1, 2 |
126 |
|
View Full Product Details