Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166116.1 | NP_001159588.1 | 3, 4 | 421 | |
NM_003275.3 | NP_003266.1 | 3, 4 | 498 | ||
GenBank mRNA | AK314533.1 | - | 3, 4 | 421 | |
BC002660.2 | - | 3, 4 | 498 | 3610014 | |
DA015088.1 | - | 3, 4 | 471 | ||
M77016.1 | - | 2, 3 | 319 |