Gene Transcripts
- Gene Symbol
- SLC16A4
- Entrez Gene ID
- 9122
- Gene Name
- solute carrier family 16 member 4
- Gene Aliases
- MCT4, MCT5
- Chromosome Location
- Chr. 1: 110362851 - 110391970 on Build GRCh38
- UniGene ID
- Hs.351306
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001201546.1 |
NP_001188475.1
|
7, 8 |
1437 |
|
NM_001201547.1 |
NP_001188476.1
|
7, 8 |
1448 |
|
NM_001201549.1 |
NP_001188478.1
|
7, 8 |
1077 |
|
NM_001319220.1 |
NP_001306149.1
|
6, 7 |
944 |
|
NM_004696.2 |
NP_004687.1
|
8, 9 |
1581 |
|
XM_005271317.4 |
XP_005271374.1
|
9, 10 |
2599 |
|
XM_006711033.3 |
XP_006711096.1
|
6, 7 |
2190 |
|
GenBank mRNA |
AK129985.1 |
- |
8, 9 |
1525 |
|
AK223307.1 |
- |
8, 9 |
1525 |
|
AK290720.1 |
- |
7, 8 |
1381 |
|
AK295946.1 |
- |
7, 8 |
1414 |
|
AK313735.1 |
- |
8, 9 |
1543 |
|
AL833619.1 |
- |
7, 8 |
1378 |
|
BC021664.1 |
- |
7, 8 |
1047 |
4050756 |
BX647948.1 |
- |
6, 7 |
910 |
|
U59185.1 |
- |
8, 9 |
1513 |
|
View Full Product Details