Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006645.2 | NP_006636.2 | 6 | 1535 | |
GenBank mRNA | AF151810.1 | - | 6 | 1071 | |
AK095668.1 | - | 6 | 1451 | ||
AK315255.1 | - | 7 | 1329 | ||
BC007919.2 | - | 7 | 1054 | 4301295 | |
BC014033.2 | - | 7 | 1054 | 4330790 | |
BC035482.1 | - | 6 | 1335 | ||
JX424571.1 | - | 2 | 40 |