Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014957.2 | NP_055772.2 | 4 | 620 | |
XM_005250838.4 | XP_005250895.2 | 4 | 511 | ||
XM_005250839.2 | XP_005250896.2 | 4 | 511 | ||
XM_005250840.4 | XP_005250897.2 | 4 | 511 | ||
XM_005250841.3 | XP_005250898.2 | 4 | 511 | ||
XM_011516933.1 | XP_011515235.1 | 4 | 511 | ||
XM_011516934.2 | XP_011515236.1 | 4 | 511 | ||
XM_011516937.1 | XP_011515239.1 | 4 | 511 | ||
XM_011516941.2 | XP_011515243.1 | 4 | 511 | ||
XM_017013241.1 | XP_016868730.1 | 4 | 513 | ||
XM_017013242.1 | XP_016868731.1 | 4 | 623 | ||
GenBank mRNA | AK058001.1 | - | 4 | 587 | |
AL544320.3 | - | 4 | 679 | ||
AW500126.1 | - | 3 | 360 | ||
BC133023.1 | - | 3 | 363 | 40147045 | |
BC144226.1 | - | 3 | 363 | 9052746 |