Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015078.3 | NP_055893.3 | 9 | 1228 | |
XM_011512585.2 | XP_011510887.1 | 1 | 670 | ||
XM_011512586.2 | XP_011510888.1 | 2 | 84 | ||
XM_017005943.1 | XP_016861432.1 | 9 | 1949 | ||
XM_017005944.1 | XP_016861433.1 | 9 | 1949 | ||
XM_017005945.1 | XP_016861434.1 | 9 | 1949 | ||
XM_017005946.1 | XP_016861435.1 | 9 | 1949 | ||
GenBank mRNA | AB020668.1 | - | 5 | 540 | |
AK124500.1 | - | 9 | 1060 | ||
AK301698.1 | - | 1 | 121 | ||
AK310962.1 | - | 9 | 1202 | ||
AY172737.1 | - | 7 | 780 | ||
BC064632.1 | - | 9 | 1044 | 5750039 | |
BX649003.1 | - | 1059 |