Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location
Chr. 14: 88385638 - 88470350 on Build GRCh38
UniGene ID
Hs.525518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040428.3 NP_001035518.1 5 474
NM_018418.4 NP_060888.2 6 570
XM_005267851.1 XP_005267908.1 6 541
XM_005267852.1 XP_005267909.1 5 470
XM_005267854.1 XP_005267911.1 7 603
XM_005267855.1 XP_005267912.1 6 536
XM_006720204.1 XP_006720267.1 6 541
XM_006720205.1 XP_006720268.1 6 541
XM_011536951.1 XP_011535253.1 6 735
XM_011536952.1 XP_011535254.1 4 353
XM_011536953.1 XP_011535255.1 5 674
XM_017021452.1 XP_016876941.1 6 735
XM_017021453.1 XP_016876942.1 6 701
XM_017021454.1 XP_016876943.1 7 603
XM_017021455.1 XP_016876944.1 6 536
XM_017021456.1 XP_016876945.1 7 549
XM_017021457.1 XP_016876946.1 5 674
GenBank mRNA AF144487.3 - 6 550
AF144488.2 - 4 275
AK055864.1 - 6 492
AK225746.1 - 7 541
AK290631.1 - 5 445
AK293007.1 - 7 523
AL136604.1 - 5 454
BC090875.1 - 6 857 30383306
CR533563.1 - 5 299
CR936777.1 - 6 516
DA568174.1 - 6 536
EU668353.1 - 6 546

View Full Product Details