Gene Transcripts

Gene Symbol
UPF3B
Entrez Gene ID
65109
Gene Name
UPF3 regulator of nonsense transcripts homolog B (yeast)
Gene Aliases
HUPF3B, MRX62, MRXS14, RENT3B, UPF3BP1, UPF3BP2, UPF3BP3, UPF3X, Upf3p-X
Chromosome Location
Chr. X: 119805311 - 119853028 on Build GRCh38
UniGene ID
Hs.103832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023010.3 NP_075386.1 1, 2 260
NM_080632.2 NP_542199.1 1, 2 260
XM_017029737.1 XP_016885226.1 1, 2 260
XM_017029738.1 XP_016885227.1 1, 2 260
XM_017029739.1 XP_016885228.1 1, 2 260
XM_017029740.1 XP_016885229.1 1, 2 260
GenBank mRNA AF318576.1 - 1, 2 180
AK093728.1 - 1, 2 198
AY013251.1 - 1, 2 230
BC121017.1 - 1, 2 203 40117102
BC121018.1 - 1, 2 203
BI549935.1 - 1, 2 240
DA949746.1 - 1, 2 260

View Full Product Details