Gene Transcripts

Gene Symbol
ERICH1
Entrez Gene ID
157697
Gene Name
glutamate rich 1
Gene Aliases
HSPC319
Chromosome Location
Chr. 8: 614737 - 731239 on Build GRCh38
UniGene ID
Hs.655310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303100.1 NP_001290029.1 4 502
NM_207332.2 NP_997215.1 4 502
XM_006716234.3 XP_006716297.2 4 491
XM_011534731.2 XP_011533033.1 4 491
XM_011534735.2 XP_011533037.1 4 491
XM_011534736.2 XP_011533038.1 4 491
XM_011534737.2 XP_011533039.1 4 491
XM_011534739.2 XP_011533041.1 4 491
XM_017013114.1 XP_016868603.1 4 491
XM_017013115.1 XP_016868604.1 4 491
XM_017013116.1 XP_016868605.1 4 491
XM_017013117.1 XP_016868606.1 4 491
XM_017013118.1 XP_016868607.1 4 491
XM_017013119.1 XP_016868608.1 4 491
XM_017013120.1 XP_016868609.1 4 491
XM_017013121.1 XP_016868610.1 4 491
XM_017013122.1 XP_016868611.1 4 491
XM_017013123.1 XP_016868612.1 4 491
XM_017013124.1 XP_016868613.1 4 491
XM_017013125.1 XP_016868614.1 4 491
XM_017013126.1 XP_016868615.1 4 491
XM_017013127.1 XP_016868616.1 4 491
XM_017013128.1 XP_016868617.1 4 491
XM_017013129.1 XP_016868618.1 4 491
XM_017013130.1 XP_016868619.1 4 491
XM_017013131.1 XP_016868620.1 4 491
XM_017013132.1 XP_016868621.1 4 491
XM_017013133.1 XP_016868622.1 4 491
XM_017013134.1 XP_016868623.1 4 491
GenBank mRNA AF161437.1 - 4 446
AK055649.1 - 3 1646
AK290264.1 - 4 486
AK297476.1 - 4 472
BC016017.1 - 523
BC046243.1 - 4 489 5207311

View Full Product Details