Gene Transcripts

Gene Symbol
TAS2R31
Entrez Gene ID
259290
Gene Name
taste 2 receptor member 31
Gene Aliases
T2R31, T2R44, T2R53, TAS2R44
Chromosome Location
Chr. 12: 11030387 - 11031407 on Build GRCh38
UniGene ID
Hs.688197
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176885.2 NP_795366.2 1 114
GenBank mRNA BC117421.1 - 1 115

View Full Product Details