Gene Transcripts

Gene Symbol
TAS2R20
Entrez Gene ID
259295
Gene Name
taste 2 receptor member 20
Gene Aliases
T2R20, T2R49, T2R56, TAS2R49
Chromosome Location
Chr. 12: 10995962 - 10997875 on Build GRCh38
UniGene ID
Hs.686384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176889.2 NP_795370.2 1 549
GenBank mRNA BC100915.2 - 1 549
BC100916.2 - 1 549 40003844
BC100917.2 - 1 549 40003847
BC100918.2 - 1 549 40003851
BC113843.1 - 1 549 40003846
BC114451.1 - 1 549 40003845

View Full Product Details