Gene Transcripts

Gene Symbol
LRRTM3
Entrez Gene ID
347731
Gene Name
leucine rich repeat transmembrane neuronal 3
Gene Aliases
Chromosome Location
Chr. 10: 66926034 - 67101551 on Build GRCh38
UniGene ID
Hs.652155
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178011.4 NP_821079.3 3 2162
NR_111909.1 3 206
GenBank mRNA AK290168.1 - 3 1695
AK294029.1 - 2 382
BC111492.1 - 3 2114 5196989
BC113715.1 - 3 1672 8322767
BC113717.1 - 3 1672 8322769
BU154601.1 - 3 189
BX640611.1 - 3 2141

View Full Product Details