Gene Transcripts

Gene Symbol
GEN1
Entrez Gene ID
348654
Gene Name
GEN1, Holliday junction 5' flap endonuclease
Gene Aliases
Gen
Chromosome Location
Chr. 2: 17753687 - 17785920 on Build GRCh38
UniGene ID
Hs.467793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130009.2 NP_001123481.2 14 2221
NM_182625.4 NP_872431.4 14 2145
XM_005262613.4 XP_005262670.1 14 2231
XM_006712005.3 XP_006712068.1 14 2128
XM_011532820.2 XP_011531122.1 14 2148
XM_011532821.2 XP_011531123.1 14 2238
XM_011532822.2 XP_011531124.1 14 2118
GenBank mRNA AK098188.1 - 7 1171
AK131387.1 - 14 2093
BC117204.1 - 13 2043 40125755
BC117206.1 - 13 2043 40125757
CR936783.1 - 14 2231

View Full Product Details