Gene Transcripts
- Gene Symbol
- DNAH12
- Entrez Gene ID
- 201625
- Gene Name
- dynein axonemal heavy chain 12
- Gene Aliases
- DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
- Chromosome Location
- Chr. 3: 57293699 - 57545203 on Build GRCh38
- UniGene ID
- Hs.201378
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291661.1 |
NP_001278590.1
|
8, 9 |
1069 |
|
NM_198564.3 |
NP_940966.2
|
8, 9 |
1069 |
|
XM_011533466.1 |
XP_011531768.1
|
8, 9 |
1066 |
|
XM_011533467.1 |
XP_011531769.1
|
8, 9 |
1033 |
|
XM_011533468.1 |
XP_011531770.1
|
8, 9 |
1066 |
|
XM_011533469.1 |
XP_011531771.1
|
8, 9 |
1066 |
|
XM_011533471.2 |
XP_011531773.1
|
8, 9 |
1066 |
|
XM_011533474.1 |
XP_011531776.1
|
8, 9 |
1066 |
|
XM_017005860.1 |
XP_016861349.1
|
8, 9 |
1066 |
|
XM_017005861.1 |
XP_016861350.1
|
8, 9 |
1928 |
|
XM_017005863.1 |
XP_016861352.1
|
8, 9 |
1066 |
|
GenBank mRNA |
AK126276.1 |
- |
8, 9 |
1066 |
|
View Full Product Details