Gene Transcripts

Gene Symbol
OR56B1
Entrez Gene ID
387748
Gene Name
olfactory receptor family 56 subfamily B member 1
Gene Aliases
OR11-65, OR56B1P
Chromosome Location
Chr. 11: 5736448 - 5737538 on Build GRCh38
UniGene ID
Hs.689609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005180.2 NP_001005180.1 1 401
GenBank mRNA AK122668.1 - 1 401
BC137184.1 - 1 398 9021181
BC137185.1 - 1 398 9021182

View Full Product Details