Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
401474
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
Chromosome Location
Chr. 8: 118131828 - 118621995 on Build GRCh38
UniGene ID
Hs.359393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101676.1 NP_001095146.1 4 465
NM_207506.2 NP_997389.2 4 465
NR_109794.1 4 380
XM_011517031.2 XP_011515333.1 4 505
XM_011517034.2 XP_011515336.1 4 504
XM_011517035.2 XP_011515337.1 4 504
XM_017013409.1 XP_016868898.1 4 477
XM_017013410.1 XP_016868899.1 4 505
XM_017013411.1 XP_016868900.1 4 475
XM_017013412.1 XP_016868901.1 4 510
GenBank mRNA AK096777.1 - 4 401
AK311704.1 - 4 380
BC121818.1 - 4 398 40109130
BC124554.1 - 4 398 40109129
BF326714.1 - 2 490
DA335826.1 - 4 465
DA459013.1 - 4 444
DB228621.1 - 4 380

View Full Product Details