Gene Transcripts

Gene Symbol
GSTM1
Entrez Gene ID
2944
Gene Name
glutathione S-transferase mu 1
Gene Aliases
GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, MU, MU-1
Chromosome Location
Chr. 1: 109687796 - 109693745 on Build GRCh38
UniGene ID
Hs.301961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000561.3 NP_000552.2 5 389
NM_146421.2 NP_666533.1 5 389
XM_005270782.4 XP_005270839.1 5 407
GenBank mRNA AY510272.1 - 5 311
AY532926.1 - 5 311
AY532927.1 - 5 311
BC024005.2 - 5 345 4823313
BC036805.1 - 5 326
BQ436476.1 - 5 375
CR541868.1 - 5 311
J03817.1 - 5 326
KJ534856.1 - 5 311
KJ534857.1 - 5 311
KJ534858.1 - 5 311
KJ534998.1 - 4 229
KJ535021.1 - 4 362
X08020.1 - 5 335

View Full Product Details