Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001174146.1 | NP_001167617.1 | 2 | 292 | |
NM_001174147.1 | NP_001167618.1 | 2 | 292 | ||
NM_002316.3 | NP_002307.2 | 2 | 292 | ||
GenBank mRNA | AF057135.1 | - | 2 | 216 | |
BC069588.1 | - | 2 | 211 | ||
BC069601.1 | - | 2 | 216 | 7262383 | |
BC112120.1 | - | 2 | 266 | 8327588 | |
BC113491.1 | - | 2 | 266 | 8322543 | |
BC143801.1 | - | 2 | 266 | 9052315 | |
KU178166.1 | - | 2 | 216 | ||
KU178167.1 | - | 2 | 216 |