Gene Transcripts

Gene Symbol
FBXL8
Entrez Gene ID
55336
Gene Name
F-box and leucine rich repeat protein 8
Gene Aliases
FBL8
Chromosome Location
Chr. 16: 67159988 - 67164174 on Build GRCh38
UniGene ID
Hs.710714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018378.2 NP_060848.2 3 393
GenBank mRNA AK002140.1 - 3 393
BC014414.1 - 3 393 4548146

View Full Product Details