Gene Transcripts
- Gene Symbol
- PRAP1
- Entrez Gene ID
- 118471
- Gene Name
- proline rich acidic protein 1
- Gene Aliases
- PRO1195, UPA
- Chromosome Location
- Chr. 10: 133347340 - 133352683 on Build GRCh38
- UniGene ID
- Hs.15951
- Species
- Homo sapiens
- Gene Symbol
- ZNF511-PRAP1
- Entrez Gene ID
- 104326056
- Gene Name
- ZNF511-PRAP1 readthrough
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.15951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EU937518.1 |
- |
5 |
523 |
|
View Full Product Details