Gene Transcripts

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
Chromosome Location
Chr. 3: 126524439 - 126558965 on Build GRCh38
UniGene ID
Hs.178210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152533.2 NP_689746.1 2 402
NR_130715.1 2 402
XM_011512455.2 XP_011510757.1 2 402
XM_011512457.2 XP_011510759.1 2 402
XM_017005751.1 XP_016861240.1 2 372
XM_017005752.1 XP_016861241.1 2 234
XM_017005753.1 XP_016861242.1 2 402
GenBank mRNA AK097841.1 - 2 402
BC032025.1 - 2 352 5165128
DB055549.1 - 2 402

View Full Product Details