Gene Transcripts
- Gene Symbol
- PABPC1P2
- Entrez Gene ID
- 728773
- Gene Name
- poly(A) binding protein cytoplasmic 1 pseudogene 2
- Gene Aliases
- PABP2, PABP4, PABPCP2, PABPCP4
- Chromosome Location
- Chr. 2: 146587057 - 146590990 on Build GRCh38
- UniGene ID
- Hs.334462
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026904.1 |
|
1 |
2666 |
|
GenBank mRNA |
BC033032.1 |
- |
1 |
2281 |
|
BC068242.1 |
- |
1 |
2030 |
|
- Gene Symbol
- GJC1
- Entrez Gene ID
- 10052
- Gene Name
- gap junction protein gamma 1
- Gene Aliases
- CX45, GJA7
- Chromosome Location
- Chr. 17: 44794988 - 44830811 on Build GRCh38
- UniGene ID
- Hs.712052
- Species
- Homo sapiens
- Gene Symbol
- LRRC57
- Entrez Gene ID
- 255252
- Gene Name
- leucine rich repeat containing 57
- Gene Aliases
- Chromosome Location
- Chr. 15: 42528332 - 42548804 on Build GRCh38
- UniGene ID
- Hs.234681
- Species
- Homo sapiens
View Full Product Details