Gene Transcripts

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
Chromosome Location
Chr. 17: 76527586 - 76555339 on Build GRCh38
UniGene ID
Hs.634380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017025013.1 XP_016880502.1 7 4939
XM_017025014.1 XP_016880503.1 7 4924
XM_017025015.1 XP_016880504.1 5 2528
GenBank mRNA AK054729.1 - 4 961
AK125617.1 - 7 2993

View Full Product Details