Gene Transcripts
- Gene Symbol
- TROAP
- Entrez Gene ID
- 10024
- Gene Name
- trophinin associated protein
- Gene Aliases
- TASTIN
- Chromosome Location
- Chr. 12: 49322135 - 49331733 on Build GRCh38
- UniGene ID
- Hs.524399
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100620.1 |
NP_001094090.1
|
2, 3 |
300 |
|
NM_001278324.1 |
NP_001265253.1
|
2, 3 |
300 |
|
NM_005480.3 |
NP_005471.3
|
2, 3 |
300 |
|
XM_006719181.1 |
XP_006719244.1
|
2, 3 |
244 |
|
XM_011537723.1 |
XP_011536025.1
|
2, 3 |
244 |
|
XM_011537725.2 |
XP_011536027.1
|
3, 4 |
436 |
|
GenBank mRNA |
AK296811.1 |
- |
2, 3 |
233 |
|
AK299302.1 |
- |
2, 3 |
205 |
|
AK315211.1 |
- |
2, 3 |
244 |
|
BC011597.2 |
- |
2, 3 |
168 |
|
BC032583.1 |
- |
2, 3 |
212 |
5498800 |
CD366215.1 |
- |
1, 2 |
568 |
|
DC410015.1 |
- |
2, 3 |
300 |
|
U04810.2 |
- |
2, 3 |
243 |
|
View Full Product Details