Gene Transcripts

Gene Symbol
RRP7A
Entrez Gene ID
27341
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
BK126B4.3, CGI-96
Chromosome Location
Chr. 22: 42508335 - 42519823 on Build GRCh38
UniGene ID
Hs.745006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015703.4 NP_056518.2 2 147
GenBank mRNA AK314650.1 - 2 113
AY039240.1 - 1 19
BC031838.1 - 1 62
BC035992.1 - 2 126
BC041639.1 - 2 105
BC042335.1 - 2 108
BC073834.1 - 2 120 5533064
BC121118.1 - 2 130 40120513
BC121119.1 - 2 130 40120514
BP314581.1 - 2 147

Gene Symbol
RRP7BP
Entrez Gene ID
91695
Gene Name
ribosomal RNA processing 7 homolog B, pseudogene
Gene Aliases
RRP7B, dJ222E13.2
Chromosome Location
Chr. 22: 42573244 - 42582011 on Build GRCh38
UniGene ID
Hs.534041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002184.2 2 120
GenBank mRNA AK098201.1 - 2 147
AL157851.1 - 2 105
BC007736.2 - 2 120
BC014647.1 - 2 118
BC143340.1 - 2 208

View Full Product Details