Gene Transcripts

Gene Symbol
RRP7A
Entrez Gene ID
27341
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
BK126B4.3, CGI-96
Chromosome Location
Chr. 22: 42508335 - 42519823 on Build GRCh38
UniGene ID
Hs.745006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015703.4 NP_056518.2 3, 4 382
GenBank mRNA AK092492.1 - 1, 2 908
AK314650.1 - 3, 4 348
AY039240.1 - 2, 3 254
BC031838.1 - 2, 3 297
BC035992.1 - 3, 4 361
BC041639.1 - 3, 4 340
BC042335.1 - 3, 4 343
BC073834.1 - 3, 4 355 5533064
BC121118.1 - 3, 4 365 40120513
BC121119.1 - 3, 4 365 40120514
BP314581.1 - 3, 4 382

Gene Symbol
RRP7BP
Entrez Gene ID
91695
Gene Name
ribosomal RNA processing 7 homolog B, pseudogene
Gene Aliases
RRP7B, dJ222E13.2
Chromosome Location
Chr. 22: 42573244 - 42582011 on Build GRCh38
UniGene ID
Hs.534041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002184.2 3, 4 348
GenBank mRNA AK074492.1 - 1, 2 109
AK098201.1 - 3, 4 375
AL157851.1 - 3, 4 333
BC007736.2 - 3, 4 348
BC014647.1 - 3, 4 346
BC143340.1 - 3, 4 436

View Full Product Details