Gene Transcripts

Gene Symbol
INHBB
Entrez Gene ID
3625
Gene Name
inhibin beta B subunit
Gene Aliases
Chromosome Location
Chr. 2: 120346143 - 120351807 on Build GRCh38
UniGene ID
Hs.1735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002193.2 NP_002184.2 1 465
GenBank mRNA BC030029.2 - 1 465
M13437.1 - 1 259
M31682.1 - 1 358

View Full Product Details