Gene Transcripts

Gene Symbol
POU6F1
Entrez Gene ID
5463
Gene Name
POU class 6 homeobox 1
Gene Aliases
BRN5, MPOU, TCFB1
Chromosome Location
Chr. 12: 51186936 - 51218134 on Build GRCh38
UniGene ID
Hs.555886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002702.3 NP_002693.3 6 1361
NR_026893.1 11 1819
XM_017019507.1 XP_016874996.1 11 3006
XM_017019508.1 XP_016874997.1 12 2917
XM_017019509.1 XP_016874998.1 10 1926
XM_017019510.1 XP_016874999.1 11 2355
XM_017019511.1 XP_016875000.1 11 2524
XM_017019512.1 XP_016875001.1 11 1880
XM_017019513.1 XP_016875002.1 11 3003
XM_017019514.1 XP_016875003.1 11 3000
XM_017019515.1 XP_016875004.1 11 2346
XM_017019516.1 XP_016875005.1 11 2997
XM_017019517.1 XP_016875006.1 10 2877
XM_017019518.1 XP_016875007.1 10 2871
XM_017019519.1 XP_016875008.1 10 2831
XM_017019520.1 XP_016875009.1 9 2233
XM_017019521.1 XP_016875010.1 9 2252
XM_017019522.1 XP_016875011.1 10 1730
XM_017019523.1 XP_016875012.1 8 1437
XM_017019524.1 XP_016875013.1 10 1828
XM_017019525.1 XP_016875014.1 11 1816
XM_017019526.1 XP_016875015.1 10 1724
XM_017019527.1 XP_016875016.1 10 1743
XM_017019528.1 XP_016875017.1 11 2249
XM_017019529.1 XP_016875018.1 10 2135
XM_017019530.1 XP_016875019.1 9 2040
GenBank mRNA AK095191.1 - 11 1819
AL832881.1 - 6 1361
BC051326.1 - 6 1164 6275848
BC074765.2 - 5 904 30915178
BC107090.1 - 1 236
L14482.1 - 6 1032
Z21966.1 - 5 1071

View Full Product Details