Gene Transcripts

Gene Symbol
GPANK1
Entrez Gene ID
7918
Gene Name
G-patch domain and ankyrin repeats 1
Gene Aliases
ANKRD59, BAT4, D6S54E, G5, GPATCH10
Chromosome Location
Chr. 6: 31661229 - 31666283 on Build GRCh38
UniGene ID
Hs.247478
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199237.1 NP_001186166.1 4 1728
NM_001199238.1 NP_001186167.1 4 1597
NM_001199239.1 NP_001186168.1 4 1594
NM_001199240.1 NP_001186169.1 4 1380
NM_033177.3 NP_149417.1 3 1704
XM_005249403.3 XP_005249460.1 4 1731
XM_006715204.1 XP_006715267.1 4 1409
XM_011514909.1 XP_011513211.1 4 1449
XM_011514910.1 XP_011513212.1 4 1446
GenBank mRNA AJ420471.1 - 3 1296
AK123707.1 - 4 1728
AK314056.1 - 4 1597
BC008783.1 - 3 1694 3629572
BC018875.2 - 3 1688

View Full Product Details