Gene Transcripts

Gene Symbol
SLC35E1
Entrez Gene ID
79939
Gene Name
solute carrier family 35 member E1
Gene Aliases
Chromosome Location
Chr. 19: 16549837 - 16572382 on Build GRCh38
UniGene ID
Hs.620596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024881.4 NP_079157.3 6 1227
GenBank mRNA AK027699.1 - 6 1021
AK027850.1 - 4 638
AK075355.1 - 4 666
BC014557.2 - 3 574
BC062562.1 - 6 1227 6598965
BX640756.1 - 6 838

View Full Product Details