Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85977935-85978430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
UniGene
Hs.163893
Gene Chromosome Location
Chr.11: 85957171-86069881 on Build GRCh38
Transcripts
NM_001008660.2 NM_001206946.1 NM_001206947.1 NM_007166.3 XM_005274322.2 XM_005274323.2 XM_005274324.2 XM_005274325.2 XM_005274326.2 XM_005274327.2 XM_005274328.2 XM_005274329.3 XM_005274330.2 XM_005274331.2 XM_005274332.2 XM_005274333.2 XM_005274334.2 XM_005274335.2 XM_005274336.2 XM_005274337.2 XM_005274340.2 XM_006718699.2 XM_006718700.2 XM_006718701.3 XM_011545293.1 XM_017018381.1 XM_017018382.1 XM_017018383.1 XM_017018384.1 XM_017018385.1 XM_017018386.1 XM_017018387.1 XM_017018388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687822 c.1639C>T p.H547Y Chr.11: 85978084-85978084 on Build GRCh38
1357533 c.1653+1G>A p.? Chr.11: 85978069-85978069 on Build GRCh38
6745932 c.1637T>C p.M546T Chr.11: 85978086-85978086 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs186913869 Chr.11: 85978336-85978336 on Build GRCh38
rs895217724 Chr.11: 85977987-85977987 on Build GRCh38
rs986544671 Chr.11: 85978136-85978136 on Build GRCh38
rs569294673 Chr.11: 85978393-85978393 on Build GRCh38
rs529145948 Chr.11: 85978247-85978247 on Build GRCh38
rs1034116602 Chr.11: 85978234-85978234 on Build GRCh38
rs978866366 Chr.11: 85978369-85978371 on Build GRCh38
rs35715728 Chr.11: 85978187-85978188 on Build GRCh38
rs375351370 Chr.11: 85978128-85978128 on Build GRCh38
rs618629 Chr.11: 85978392-85978392 on Build GRCh38
rs952569945 Chr.11: 85978201-85978204 on Build GRCh38
rs774398467 Chr.11: 85978054-85978054 on Build GRCh38
rs957275119 Chr.11: 85978326-85978326 on Build GRCh38
rs372053430 Chr.11: 85978070-85978070 on Build GRCh38
rs377457728 Chr.11: 85978266-85978266 on Build GRCh38
rs572016034 Chr.11: 85978256-85978256 on Build GRCh38
rs1031585172 Chr.11: 85978317-85978317 on Build GRCh38
rs939669480 Chr.11: 85978160-85978160 on Build GRCh38
rs775534184 Chr.11: 85978040-85978040 on Build GRCh38
rs921044220 Chr.11: 85978110-85978110 on Build GRCh38
rs544181592 Chr.11: 85978391-85978391 on Build GRCh38
rs927194933 Chr.11: 85977957-85977957 on Build GRCh38
rs1019280664 Chr.11: 85978171-85978172 on Build GRCh38
rs866011893 Chr.11: 85978016-85978016 on Build GRCh38
rs868095153 Chr.11: 85978331-85978331 on Build GRCh38
rs371402533 Chr.11: 85978050-85978050 on Build GRCh38
rs111447339 Chr.11: 85978207-85978207 on Build GRCh38
rs773159747 Chr.11: 85978069-85978069 on Build GRCh38
rs200457806 Chr.11: 85978025-85978025 on Build GRCh38
rs749493839 Chr.11: 85978042-85978042 on Build GRCh38
rs1013610221 Chr.11: 85977994-85977994 on Build GRCh38
rs618639 Chr.11: 85978385-85978385 on Build GRCh38
rs138593326 Chr.11: 85978137-85978137 on Build GRCh38
rs926015950 Chr.11: 85978373-85978373 on Build GRCh38
rs781765785 Chr.11: 85978033-85978033 on Build GRCh38
rs199558359 Chr.11: 85978132-85978132 on Build GRCh38
rs141993999 Chr.11: 85978088-85978088 on Build GRCh38
rs566864027 Chr.11: 85978007-85978007 on Build GRCh38
rs752188373 Chr.11: 85978123-85978123 on Build GRCh38
rs938709817 Chr.11: 85977966-85977966 on Build GRCh38
rs577876060 Chr.11: 85978205-85978205 on Build GRCh38
rs946028982 Chr.11: 85978360-85978360 on Build GRCh38
rs1026688175 Chr.11: 85978196-85978196 on Build GRCh38
rs1009541161 Chr.11: 85978168-85978169 on Build GRCh38
rs183128872 Chr.11: 85978171-85978171 on Build GRCh38
rs766169180 Chr.11: 85978077-85978077 on Build GRCh38
rs193085915 Chr.11: 85978060-85978060 on Build GRCh38
rs966704343 Chr.11: 85978009-85978009 on Build GRCh38
rs754647684 Chr.11: 85978101-85978101 on Build GRCh38
rs1055280904 Chr.11: 85978209-85978209 on Build GRCh38
rs757544221 Chr.11: 85978077-85978077 on Build GRCh38
rs369526946 Chr.11: 85978129-85978129 on Build GRCh38
rs770215660 Chr.11: 85978038-85978038 on Build GRCh38
rs1037325154 Chr.11: 85978161-85978161 on Build GRCh38
rs765051345 Chr.11: 85978121-85978121 on Build GRCh38
rs1046440152 Chr.11: 85978261-85978261 on Build GRCh38
rs778718868 Chr.11: 85978063-85978063 on Build GRCh38
rs1028131893 Chr.11: 85978370-85978370 on Build GRCh38
rs779396750 Chr.11: 85978101-85978102 on Build GRCh38
rs562073702 Chr.11: 85978348-85978348 on Build GRCh38
rs553539230 Chr.11: 85978253-85978253 on Build GRCh38
rs747939914 Chr.11: 85978103-85978103 on Build GRCh38
rs1011390382 Chr.11: 85978369-85978369 on Build GRCh38
rs890726125 Chr.11: 85978133-85978133 on Build GRCh38
rs370802160 Chr.11: 85978175-85978175 on Build GRCh38
rs746162206 Chr.11: 85978036-85978036 on Build GRCh38
rs1057143785 Chr.11: 85977979-85977982 on Build GRCh38
rs999532235 Chr.11: 85978313-85978313 on Build GRCh38

View Full Product Details