Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113399747-113400261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
UniGene
Hs.448473
Gene Chromosome Location
Chr.11: 113386014-113400418 on Build GRCh38
Transcripts
NM_178510.1 XM_011542736.2 XM_011542737.2 XM_011542738.2 XM_017017475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6045697 c.1812C>T p.G604G Chr.11: 113399781-113399781 on Build GRCh38
79495 c.2037A>T p.L679L Chr.11: 113400006-113400006 on Build GRCh38
3443515 c.1922T>A p.V641E Chr.11: 113399891-113399891 on Build GRCh38
6130903 c.1975G>T p.G659C Chr.11: 113399944-113399944 on Build GRCh38
3443517 c.2059G>A p.A687T Chr.11: 113400028-113400028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375843671 Chr.11: 113400167-113400167 on Build GRCh38
rs961308840 Chr.11: 113400009-113400009 on Build GRCh38
rs766436729 Chr.11: 113399983-113399983 on Build GRCh38
rs375251188 Chr.11: 113399876-113399876 on Build GRCh38
rs953986849 Chr.11: 113399775-113399775 on Build GRCh38
rs769529971 Chr.11: 113399834-113399834 on Build GRCh38
rs869040191 Chr.11: 113399976-113399976 on Build GRCh38
rs751601411 Chr.11: 113400048-113400048 on Build GRCh38
rs201092692 Chr.11: 113399978-113399978 on Build GRCh38
rs980219848 Chr.11: 113400035-113400035 on Build GRCh38
rs757386062 Chr.11: 113399797-113399797 on Build GRCh38
rs750102788 Chr.11: 113400062-113400062 on Build GRCh38
rs146480879 Chr.11: 113400055-113400055 on Build GRCh38
rs768518656 Chr.11: 113400077-113400077 on Build GRCh38
rs750469108 Chr.11: 113399886-113399886 on Build GRCh38
rs772721743 Chr.11: 113399839-113399839 on Build GRCh38
rs77077463 Chr.11: 113400027-113400027 on Build GRCh38
rs55849504 Chr.11: 113399927-113399927 on Build GRCh38
rs773111950 Chr.11: 113400007-113400007 on Build GRCh38
rs748104280 Chr.11: 113400164-113400164 on Build GRCh38
rs377621768 Chr.11: 113400032-113400032 on Build GRCh38
rs1800497 Chr.11: 113400106-113400106 on Build GRCh38
rs139904975 Chr.11: 113400183-113400183 on Build GRCh38
rs933133026 Chr.11: 113400042-113400042 on Build GRCh38
rs748945337 Chr.11: 113399921-113399921 on Build GRCh38
rs772170574 Chr.11: 113400202-113400202 on Build GRCh38
rs775420902 Chr.11: 113399875-113399875 on Build GRCh38
rs918149045 Chr.11: 113400068-113400068 on Build GRCh38
rs958033283 Chr.11: 113400173-113400173 on Build GRCh38
rs774363217 Chr.11: 113400108-113400108 on Build GRCh38
rs371927986 Chr.11: 113400151-113400151 on Build GRCh38
rs374434021 Chr.11: 113400067-113400067 on Build GRCh38
rs945343280 Chr.11: 113399801-113399801 on Build GRCh38
rs371542276 Chr.11: 113400126-113400126 on Build GRCh38
rs765409298 Chr.11: 113399881-113399881 on Build GRCh38
rs370756598 Chr.11: 113400030-113400030 on Build GRCh38
rs528190275 Chr.11: 113399889-113399889 on Build GRCh38
rs770983683 Chr.11: 113400080-113400080 on Build GRCh38
rs371374955 Chr.11: 113400088-113400088 on Build GRCh38
rs776925300 Chr.11: 113400223-113400223 on Build GRCh38
rs764424497 Chr.11: 113399877-113399877 on Build GRCh38
rs775862159 Chr.11: 113400232-113400232 on Build GRCh38
rs369403733 Chr.11: 113400146-113400146 on Build GRCh38
rs192587080 Chr.11: 113400031-113400031 on Build GRCh38
rs150627679 Chr.11: 113399873-113399873 on Build GRCh38
rs766047837 Chr.11: 113400145-113400145 on Build GRCh38
rs781581458 Chr.11: 113399920-113399920 on Build GRCh38
rs866128139 Chr.11: 113399957-113399957 on Build GRCh38
rs749107081 Chr.11: 113400170-113400170 on Build GRCh38
rs534472707 Chr.11: 113400216-113400216 on Build GRCh38
rs56006094 Chr.11: 113399977-113399977 on Build GRCh38
rs1013702069 Chr.11: 113400116-113400116 on Build GRCh38
rs759484490 Chr.11: 113399872-113399872 on Build GRCh38
rs781258690 Chr.11: 113399771-113399771 on Build GRCh38
rs139195339 Chr.11: 113399888-113399888 on Build GRCh38
rs79635225 Chr.11: 113400144-113400144 on Build GRCh38
rs376599631 Chr.11: 113400176-113400176 on Build GRCh38
rs760087338 Chr.11: 113400147-113400147 on Build GRCh38
rs368972146 Chr.11: 113400084-113400084 on Build GRCh38
rs978011371 Chr.11: 113399812-113399812 on Build GRCh38
rs373202169 Chr.11: 113400022-113400022 on Build GRCh38
rs200344768 Chr.11: 113400042-113400042 on Build GRCh38
rs755322242 Chr.11: 113399912-113399912 on Build GRCh38
rs779573357 Chr.11: 113400025-113400025 on Build GRCh38
rs760609097 Chr.11: 113399946-113399946 on Build GRCh38
rs533178221 Chr.11: 113399999-113399999 on Build GRCh38
rs1051551088 Chr.11: 113400052-113400052 on Build GRCh38
rs768579058 Chr.11: 113400028-113400028 on Build GRCh38
rs987084811 Chr.11: 113399798-113399798 on Build GRCh38
rs775266427 Chr.11: 113399941-113399941 on Build GRCh38
rs202222056 Chr.11: 113399938-113399938 on Build GRCh38
rs779083285 Chr.11: 113399817-113399817 on Build GRCh38
rs758905204 Chr.11: 113399823-113399823 on Build GRCh38
rs768363214 Chr.11: 113399962-113399962 on Build GRCh38
rs200745528 Chr.11: 113400001-113400001 on Build GRCh38
rs773076411 Chr.11: 113400089-113400089 on Build GRCh38
rs17115461 Chr.11: 113400006-113400006 on Build GRCh38
rs775477797 Chr.11: 113400215-113400215 on Build GRCh38
rs758040982 Chr.11: 113400143-113400143 on Build GRCh38
rs775999058 Chr.11: 113400115-113400115 on Build GRCh38
rs778061889 Chr.11: 113400056-113400056 on Build GRCh38
rs376195155 Chr.11: 113399778-113399778 on Build GRCh38
rs565700593 Chr.11: 113399878-113399878 on Build GRCh38
rs763198483 Chr.11: 113400040-113400040 on Build GRCh38
rs745375509 Chr.11: 113399937-113399937 on Build GRCh38
rs760424591 Chr.11: 113399788-113399789 on Build GRCh38
rs1056605004 Chr.11: 113399896-113399896 on Build GRCh38
rs746405597 Chr.11: 113399818-113399818 on Build GRCh38
rs777770244 Chr.11: 113400169-113400169 on Build GRCh38
rs368854368 Chr.11: 113400015-113400015 on Build GRCh38
rs756414586 Chr.11: 113399768-113399768 on Build GRCh38
rs771504640 Chr.11: 113400019-113400019 on Build GRCh38
rs770514908 Chr.11: 113399923-113399923 on Build GRCh38
rs1039293894 Chr.11: 113399859-113399859 on Build GRCh38
rs764568482 Chr.11: 113400219-113400219 on Build GRCh38
rs938360716 Chr.11: 113399894-113399894 on Build GRCh38
rs747460465 Chr.11: 113399910-113399910 on Build GRCh38
rs762136964 Chr.11: 113399975-113399975 on Build GRCh38
rs760021366 Chr.11: 113400051-113400051 on Build GRCh38
rs749486188 Chr.11: 113400105-113400105 on Build GRCh38
rs374980288 Chr.11: 113400142-113400142 on Build GRCh38
rs189187004 Chr.11: 113399967-113399967 on Build GRCh38
rs748891836 Chr.11: 113399844-113399844 on Build GRCh38
rs903941492 Chr.11: 113399969-113399969 on Build GRCh38
rs770459701 Chr.11: 113399852-113399852 on Build GRCh38
rs889896236 Chr.11: 113400117-113400117 on Build GRCh38
rs773785910 Chr.11: 113399857-113399857 on Build GRCh38
rs201580434 Chr.11: 113400175-113400175 on Build GRCh38
rs757998438 Chr.11: 113400066-113400066 on Build GRCh38
rs367992103 Chr.11: 113400114-113400114 on Build GRCh38
rs373359910 Chr.11: 113400161-113400161 on Build GRCh38
rs780529370 Chr.11: 113399827-113399827 on Build GRCh38
rs780396307 Chr.11: 113399895-113399895 on Build GRCh38
rs753990582 Chr.11: 113399787-113399787 on Build GRCh38
rs778503691 Chr.11: 113400016-113400016 on Build GRCh38
rs564355960 Chr.11: 113399985-113399985 on Build GRCh38
rs774527374 Chr.11: 113399878-113399878 on Build GRCh38
rs780019234 Chr.11: 113399874-113399874 on Build GRCh38
rs201856441 Chr.11: 113399833-113399833 on Build GRCh38
rs746512232 Chr.11: 113400069-113400069 on Build GRCh38
rs866097984 Chr.11: 113399800-113399800 on Build GRCh38
rs773594486 Chr.11: 113400039-113400039 on Build GRCh38
rs753097769 Chr.11: 113400054-113400054 on Build GRCh38

View Full Product Details