Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21294682-21295215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
UniGene
Hs.126035
Gene Chromosome Location
Chr.14: 21287977-21351316 on Build GRCh38
Transcripts
NM_020366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
954370 c.160C>T p.R54C Chr.14: 21294751-21294751 on Build GRCh38
6200215 c.156A>C p.R52R Chr.14: 21294747-21294747 on Build GRCh38
5671077 c.126C>A p.N42K Chr.14: 21294717-21294717 on Build GRCh38
344273 c.191C>T p.S64F Chr.14: 21294782-21294782 on Build GRCh38
6075357 c.110C>T p.P37L Chr.14: 21294701-21294701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199606938 Chr.14: 21294744-21294744 on Build GRCh38
rs1053716484 Chr.14: 21295093-21295093 on Build GRCh38
rs754849037 Chr.14: 21294755-21294755 on Build GRCh38
rs756243895 Chr.14: 21295023-21295023 on Build GRCh38
rs569328502 Chr.14: 21294992-21294992 on Build GRCh38
rs1054601338 Chr.14: 21295179-21295179 on Build GRCh38
rs544775892 Chr.14: 21295170-21295170 on Build GRCh38
rs773027748 Chr.14: 21294847-21294847 on Build GRCh38
rs566585402 Chr.14: 21294868-21294868 on Build GRCh38
rs772414010 Chr.14: 21294831-21294842 on Build GRCh38
rs1029614017 Chr.14: 21295041-21295041 on Build GRCh38
rs553115940 Chr.14: 21295096-21295096 on Build GRCh38
rs918235158 Chr.14: 21295115-21295115 on Build GRCh38
rs752476122 Chr.14: 21294776-21294776 on Build GRCh38
rs746359185 Chr.14: 21294831-21294839 on Build GRCh38
rs375828077 Chr.14: 21294947-21294947 on Build GRCh38
rs1034300979 Chr.14: 21295146-21295146 on Build GRCh38
rs757352536 Chr.14: 21294798-21294798 on Build GRCh38
rs760608252 Chr.14: 21294831-21294837 on Build GRCh38
rs569460428 Chr.14: 21294953-21294953 on Build GRCh38
rs998496113 Chr.14: 21295032-21295032 on Build GRCh38
rs62646878 Chr.14: 21294786-21294786 on Build GRCh38
rs557889188 Chr.14: 21295139-21295139 on Build GRCh38
rs755845899 Chr.14: 21294778-21294778 on Build GRCh38
rs771414067 Chr.14: 21294831-21294841 on Build GRCh38
rs919462616 Chr.14: 21294909-21294909 on Build GRCh38
rs112381809 Chr.14: 21295053-21295053 on Build GRCh38
rs552781902 Chr.14: 21294984-21294984 on Build GRCh38
rs750443729 Chr.14: 21295006-21295006 on Build GRCh38
rs532459775 Chr.14: 21294904-21294904 on Build GRCh38
rs370643056 Chr.14: 21294815-21294815 on Build GRCh38
rs376250340 Chr.14: 21294752-21294752 on Build GRCh38
rs370209211 Chr.14: 21295194-21295194 on Build GRCh38
rs927531129 Chr.14: 21295140-21295140 on Build GRCh38
rs914760508 Chr.14: 21295180-21295180 on Build GRCh38
rs376435824 Chr.14: 21294707-21294707 on Build GRCh38
rs749473223 Chr.14: 21294795-21294795 on Build GRCh38
rs909674828 Chr.14: 21294889-21294889 on Build GRCh38
rs879141269 Chr.14: 21294937-21294937 on Build GRCh38
rs137853124 Chr.14: 21294785-21294785 on Build GRCh38
rs972481656 Chr.14: 21295111-21295111 on Build GRCh38
rs374171080 Chr.14: 21294746-21294746 on Build GRCh38
rs765765074 Chr.14: 21294753-21294753 on Build GRCh38
rs192003551 Chr.14: 21294745-21294745 on Build GRCh38
rs906642621 Chr.14: 21295126-21295126 on Build GRCh38
rs779599668 Chr.14: 21294831-21294840 on Build GRCh38
rs139211878 Chr.14: 21295108-21295108 on Build GRCh38
rs185309465 Chr.14: 21295031-21295031 on Build GRCh38
rs1016730071 Chr.14: 21295095-21295095 on Build GRCh38
rs754676113 Chr.14: 21294993-21294993 on Build GRCh38
rs941115329 Chr.14: 21294931-21294931 on Build GRCh38
rs982392566 Chr.14: 21294975-21294975 on Build GRCh38
rs34477171 Chr.14: 21294808-21294809 on Build GRCh38
rs1054631928 Chr.14: 21295001-21295001 on Build GRCh38
rs973677200 Chr.14: 21294869-21294869 on Build GRCh38
rs772456915 Chr.14: 21294818-21294818 on Build GRCh38
rs1009589542 Chr.14: 21295114-21295114 on Build GRCh38
rs921104599 Chr.14: 21295052-21295052 on Build GRCh38
rs976713441 Chr.14: 21294775-21294775 on Build GRCh38
rs770933786 Chr.14: 21294732-21294732 on Build GRCh38
rs768072319 Chr.14: 21294940-21294940 on Build GRCh38
rs985219728 Chr.14: 21295089-21295089 on Build GRCh38
rs777888377 Chr.14: 21294727-21294727 on Build GRCh38
rs926280334 Chr.14: 21295087-21295087 on Build GRCh38
rs530008238 Chr.14: 21294754-21294754 on Build GRCh38
rs546692544 Chr.14: 21294810-21294810 on Build GRCh38
rs769072975 Chr.14: 21294842-21294842 on Build GRCh38
rs751975588 Chr.14: 21294718-21294718 on Build GRCh38
rs148321752 Chr.14: 21295014-21295014 on Build GRCh38
rs939654965 Chr.14: 21295125-21295125 on Build GRCh38
rs950331071 Chr.14: 21294959-21294959 on Build GRCh38
rs918278617 Chr.14: 21294956-21294956 on Build GRCh38
rs536209717 Chr.14: 21295064-21295064 on Build GRCh38
rs777977490 Chr.14: 21294780-21294780 on Build GRCh38
rs774956493 Chr.14: 21294736-21294736 on Build GRCh38
rs867196845 Chr.14: 21294828-21294828 on Build GRCh38
rs747388634 Chr.14: 21294826-21294826 on Build GRCh38
rs764967308 Chr.14: 21294748-21294748 on Build GRCh38
rs1020272100 Chr.14: 21295024-21295024 on Build GRCh38
rs775674497 Chr.14: 21294831-21294838 on Build GRCh38
rs200225522 Chr.14: 21294822-21294822 on Build GRCh38
rs371044065 Chr.14: 21294728-21294728 on Build GRCh38
rs781081217 Chr.14: 21294761-21294761 on Build GRCh38

View Full Product Details