Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34231048-34231568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

SNP Detail

SNP ID Chromosome Location
rs990618951 Chr.15: 34231110-34231110 on Build GRCh38
rs904762982 Chr.15: 34231255-34231255 on Build GRCh38
rs958752052 Chr.15: 34231108-34231108 on Build GRCh38
rs748128930 Chr.15: 34231363-34231363 on Build GRCh38
rs927402696 Chr.15: 34231088-34231088 on Build GRCh38
rs767668808 Chr.15: 34231302-34231302 on Build GRCh38
rs977235935 Chr.15: 34231193-34231193 on Build GRCh38
rs1034276885 Chr.15: 34231288-34231288 on Build GRCh38
rs576428619 Chr.15: 34231511-34231511 on Build GRCh38
rs781763306 Chr.15: 34231239-34231239 on Build GRCh38
rs890860839 Chr.15: 34231262-34231262 on Build GRCh38
rs1013280827 Chr.15: 34231326-34231326 on Build GRCh38
rs992869097 Chr.15: 34231303-34231303 on Build GRCh38
rs538475109 Chr.15: 34231364-34231365 on Build GRCh38
rs1047630827 Chr.15: 34231147-34231147 on Build GRCh38
rs983796369 Chr.15: 34231468-34231468 on Build GRCh38
rs1037346517 Chr.15: 34231155-34231155 on Build GRCh38
rs886051040 Chr.15: 34231197-34231197 on Build GRCh38
rs898038131 Chr.15: 34231343-34231343 on Build GRCh38
rs142451941 Chr.15: 34231222-34231222 on Build GRCh38
rs995887249 Chr.15: 34231197-34231197 on Build GRCh38
rs968736998 Chr.15: 34231073-34231073 on Build GRCh38
rs1025260021 Chr.15: 34231316-34231316 on Build GRCh38
rs59091709 Chr.15: 34231077-34231077 on Build GRCh38
rs941897588 Chr.15: 34231485-34231485 on Build GRCh38
rs756601488 Chr.15: 34231344-34231345 on Build GRCh38
rs191826889 Chr.15: 34231358-34231358 on Build GRCh38
rs941658071 Chr.15: 34231151-34231151 on Build GRCh38
rs71464853 Chr.15: 34231179-34231179 on Build GRCh38
rs1051219623 Chr.15: 34231252-34231252 on Build GRCh38
rs917718556 Chr.15: 34231123-34231123 on Build GRCh38
rs869047252 Chr.15: 34231365-34231365 on Build GRCh38
rs148928495 Chr.15: 34231512-34231512 on Build GRCh38
rs59712831 Chr.15: 34231244-34231244 on Build GRCh38
rs571069456 Chr.15: 34231159-34231159 on Build GRCh38
rs189869220 Chr.15: 34231287-34231287 on Build GRCh38
rs570028839 Chr.15: 34231191-34231191 on Build GRCh38
rs970573679 Chr.15: 34231432-34231432 on Build GRCh38
rs547947247 Chr.15: 34231521-34231523 on Build GRCh38
rs188296648 Chr.15: 34231535-34231535 on Build GRCh38
rs534786265 Chr.15: 34231357-34231357 on Build GRCh38
rs1012898007 Chr.15: 34231368-34231368 on Build GRCh38
rs199752906 Chr.15: 34231365-34231365 on Build GRCh38
rs879163897 Chr.15: 34231366-34231366 on Build GRCh38
rs949366523 Chr.15: 34231127-34231127 on Build GRCh38
rs1036480805 Chr.15: 34231342-34231342 on Build GRCh38
rs138724264 Chr.15: 34231477-34231477 on Build GRCh38
rs77227436 Chr.15: 34231377-34231378 on Build GRCh38
rs971461585 Chr.15: 34231411-34231411 on Build GRCh38
rs567355546 Chr.15: 34231254-34231254 on Build GRCh38
rs780985224 Chr.15: 34231363-34231364 on Build GRCh38
rs924470481 Chr.15: 34231208-34231208 on Build GRCh38
rs1028067333 Chr.15: 34231075-34231075 on Build GRCh38
rs183994831 Chr.15: 34231508-34231508 on Build GRCh38
rs531551756 Chr.15: 34231150-34231150 on Build GRCh38
rs1031761101 Chr.15: 34231437-34231437 on Build GRCh38
rs958782667 Chr.15: 34231301-34231301 on Build GRCh38
rs935897902 Chr.15: 34231220-34231220 on Build GRCh38
rs1024752419 Chr.15: 34231409-34231409 on Build GRCh38
rs572099278 Chr.15: 34231461-34231461 on Build GRCh38
rs921185811 Chr.15: 34231235-34231235 on Build GRCh38

View Full Product Details