Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
Chromosome Location
Chr.X: 2903970 - 2929375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001669.3 NP_001660.2 9 - 10 1523 81
XM_005274514.2 XP_005274571.1 8 - 9 1358 81
GenBank mRNA AK314479.1 - 9 - 10 1487 81
BC020229.1 - - 1451 81
X83572.1 - 9 - 10 1499 81

View Full Product Details