The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002928.3 | NP_002919.3 | 1 - 2 | 202 | 108 |
GenBank mRNA | AK301261.1 | - | 1 - 2 | 178 | 108 |
AK311880.1 | - | 1 - 2 | 178 | 108 | |
AW193671.1 | - | 1 - 2 | 197 | 108 | |
BC006243.2 | - | 1 - 2 | 164 | 108 | |
DC349953.1 | - | 1 - 2 | 206 | 108 | |
U70426.1 | - | 1 - 2 | 140 | 108 |