The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308248.1 | NP_001295177.1 | 1 - 2 | 67 | 64 |
NM_005665.5 | NP_005656.4 | 1 - 2 | 67 | 64 | |
XM_017002269.1 | XP_016857758.1 | 1 - 2 | 65 | 64 | |
XM_017002271.1 | XP_016857760.1 | 1 - 2 | 65 | 64 | |
XM_017002272.1 | XP_016857761.1 | 1 - 2 | 65 | 64 | |
XM_017002277.1 | XP_016857766.1 | 1 - 2 | 65 | 64 | |
XM_017002278.1 | XP_016857767.1 | 1 - 2 | 65 | 64 | |
XM_017002282.1 | XP_016857771.1 | 1 - 2 | 65 | 64 | |
XM_017002283.1 | XP_016857772.1 | 1 - 2 | 65 | 64 | |
GenBank mRNA | AB449875.1 | - | 1 - 2 | 57 | 64 |
AF008915.1 | - | 1 - 2 | 68 | 64 | |
BP282704.1 | - | 1 - 2 | 67 | 64 |