The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000388.3 | NP_000379.2 | 1 - 2 | 133 | 70 |
GenBank mRNA | U20759.1 | - | 1 - 2 | 133 | 70 |