The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000590.1 | NP_000581.1 | 3 - 4 | 199 | 92 |
GenBank mRNA | BC066283.1 | - | 3 - 4 | 199 | 92 |
BC066284.1 | - | 3 - 4 | 199 | 92 | |
BC066285.1 | - | 3 - 4 | 199 | 92 | |
BC066286.1 | - | 3 - 4 | 199 | 92 | |
M30134.1 | - | 3 - 4 | 199 | 92 | |
S63356.1 | - | 3 - 4 | 199 | 92 | |
X17543.1 | - | 3 - 4 | 199 | 92 |