The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003565.2 | NP_003556.1 | 26 - 27 | 3230 | 78 |
XM_011538798.2 | XP_011537100.1 | 26 - 27 | 3076 | 78 | |
XM_011538799.2 | XP_011537101.1 | 26 - 27 | 2989 | 78 | |
GenBank mRNA | AB018265.2 | - | 26 - 27 | 3012 | 78 |
AF045458.1 | - | 26 - 27 | 3230 | 78 |